Learn More
Description
Specifications
Specifications
| For Use With (Application) | Enzyme Assay Substrate |
| Name | Collagen (Bovine) |
| Preparation Method | Native protein purified from Bovine Achilles Tendon according to the method of Einbinder and Schubert, J. Biol. Chem., 188, 335.(1951) |
| Quantity | 1 g |
| Storage Requirements | Store at 4°C. This product is stable for at least 3 months. |
| Regulatory Status | RUO |
| Common Name | COLLAGEN |
| Species | Bovine |
| Protein Tag | None |
| Expression System | Native protein purified from Bovine Achilles Tendon according to the method of Einbinder and Schubert, J. Biol. Chem., 188, 335.(1951) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.