Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen, Bovine Achilles tendon

Catalog No. AAJ6021809
Change view
Click to view available options
Quantity:
1g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6021809 10g
AAJ6021803 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6021809 Supplier Thermo Scientific Chemicals Supplier No. J6021809
Add to Cart

missing translation for 'validMainframeMsg'

For research use

Collagen, bovine achilles tendon is suitable for cell adhesion in tissue culture, as well as gel formation, platelet aggregation, and as a substrate for collagenase. It is also been used as cell culture substratum, adhesion. It is also used as an attachment factor for primary cultures of epithelioid cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Attachment and Proliferation
Formulation Lyophilized powder
Quantity 10g
Source Bovine achilles tendon
Storage Requirements 2° to 8°C
Regulatory Status RUO
Product Type Collagen, Bovine achilles tendon
Recombinant Native
Form Lyophilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.