Learn More
Sigma Aldrich Fine Chemicals Biosciences Collagen calf skin Bo100MG
Catalog No. 502891509
Encompass
Collagen type I is a component of skin, bone, tendon, and other fibrous connective tissues. It is a left handed helix with three polypeptide chains and contains repeating units of glycine, proline and hydroxyproline amino acids. It is a component of extracellular matrix and close to 28 types is present in bovine.
Catalog No. 50-289-1509 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C9791100MG
Collagen type I is a component of skin, bone, tendon, and other fibrous connective tissues. It is a left handed helix with three polypeptide chains and contains repeating units of glycine, proline and hydroxyproline amino acids. It is a component of extracellular matrix and close to 28 types is present in bovine.