Learn More
Advanced Biomatrix 48-well Plates, PureCol Collagen Coated
Catalog No. 501960288
Encompass_Preferred
PureCol Collagen Coated, 48-well Plates with a flat bottom have a uniform and consistent application of high quality Type I collagen on clear polystyrene surface. Plates are compatible for use with all common dispensers, readers and washers. Plates are aseptically processed and are non-pyrogenic. Plates are available in quantities of 5 plates per sleeve.
Catalog No. 50-196-0288 Supplier Advanced Biomatrix Supplier No. 51815EA
PureCol Collagen Coated, 48-well Plates with a flat bottom have a uniform and consistent application of high quality Type I collagen on clear polystyrene surface. Plates are compatible for use with all common dispensers, readers and washers. Plates are aseptically processed and are non-pyrogenic. Plates are available in quantities of 5 plates per sleeve.