Learn More
Advanced Biomatrix 6-well Plates, PureCol Collagen Coated
Catalog No. 501960276
Encompass_Preferred
PureCol Collagen Coated, 6-well Plates with a flat bottom have a uniform and consistent application of high quality Type I collagen on clear polystyrene surface. Plates are compatible for use with all common dispensers, readers and washers. Plates are aseptically processed and are non-pyrogenic. Plates are available in quantities of 5 plates per sleeve.
Catalog No. 50-196-0276 Supplier Advanced Biomatrix Supplier No. 50735EA
PureCol Collagen Coated, 6-well Plates with a flat bottom have a uniform and consistent application of high quality Type I collagen on clear polystyrene surface. Plates are compatible for use with all common dispensers, readers and washers. Plates are aseptically processed and are non-pyrogenic. Plates are available in quantities of 5 plates per sleeve.