Learn More
Advanced Biomatrix 96-well Plates, PureCol Collagen Coated
Catalog No. 501960275
Encompass_Preferred
PureCol Collagen Coated, 96-well Plates with a flat bottom have an uniform and consistent application of high quality Type I collagen on clear polystyrene surface. Plates are compatible for use with all common dispensers, readers and washers. Plates are aseptically processed and are non-pyrogenic. Plates are available in quantities of 5 plates per sleeve.
Catalog No. 50-196-0275 Supplier Advanced Biomatrix Supplier No. 50725EA
PureCol Collagen Coated, 96-well Plates with a flat bottom have an uniform and consistent application of high quality Type I collagen on clear polystyrene surface. Plates are compatible for use with all common dispensers, readers and washers. Plates are aseptically processed and are non-pyrogenic. Plates are available in quantities of 5 plates per sleeve.