Learn More
Sigma Aldrich Fine Chemicals Biosciences Collagen from bovine achilles tendon powder, suitable for substrate for collagenase | 9007-34-5 | MFCD00130825 | 5G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C98795G
Encompass
Collagen from bovine achilles tendon powder, suitable for substrate for collagenase | 9007-34-5 | MFCD00130825 | 5G
Catalog No. 50-178-1036