Learn More
Sigma Aldrich Fine Chemicals Biosciences Collagen from bovine flexo
Catalog No. 502901206
Encompass
This product is supplied as a lyophilized fibrous powder in a 1 gram package size. Bioburden and endotoxin levels are tested - this product is not considered sterile.This collagen product is naturally cross-linked yielding a robust material for applications which require structure and strength. This product can be readily prepared into such forms as tissue scaffolds foams sponges suspensions coatings putties films and sheets but does not form hydrogels. Using typical cross-linking methods this material can be tuned for optimal in vivo resorption. This collagen product is ideal for tissue engineering applications and uses with inorganic and biomaterials. The product is provided in user-friendly packaging for use and storage. Avoid extended exposure to ambient environment since this material is hygroscopic.
Catalog No. 50-290-1206 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 51621G
This product is supplied as a lyophilized fibrous powder in a 1 gram package size. Bioburden and endotoxin levels are tested - this product is not considered sterile.This collagen product is naturally cross-linked yielding a robust material for applications which require structure and strength. This product can be readily prepared into such forms as tissue scaffolds foams sponges suspensions coatings putties films and sheets but does not form hydrogels. Using typical cross-linking methods this material can be tuned for optimal in vivo resorption. This collagen product is ideal for tissue engineering applications and uses with inorganic and biomaterials. The product is provided in user-friendly packaging for use and storage. Avoid extended exposure to ambient environment since this material is hygroscopic.