Learn More
Sigma Aldrich Fine Chemicals Biosciences Collagen from rat tail ten
Catalog No. 502905384
Encompass
Collagens are abundantly present in vertebrates. 41 specific genes codes for 42 polypeptide chains and produce 27 collagen types. Collagens nbspare mainly present in the extracellular matrix. Collagen type I is abundantly present in mammals. Type IV collagen is an important structural protein of basement membranes. It is chemically and genetically different from stroma collagen types I and III and cartilage collagen type II.
Catalog No. 50-290-5384 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11179179001
Collagens are abundantly present in vertebrates. 41 specific genes codes for 42 polypeptide chains and produce 27 collagen types. Collagens nbspare mainly present in the extracellular matrix. Collagen type I is abundantly present in mammals. Type IV collagen is an important structural protein of basement membranes. It is chemically and genetically different from stroma collagen types I and III and cartilage collagen type II.