Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Collagen I, Coated Plate, 6 well

Catalog No. A1142801
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1142801 5 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1142801 Supplier Gibco™ Supplier No. A1142801
  • $144.00
    $132.65 / Each of 1

    Save $11.35

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Collagen I, Coated Plate, 6 well

Collagen I, Rat Tail is the cell culture workhorse of basic coating and gel preparations for growing attachment cells. Gibco™ Collagen I, Rat Tail is now available in pre-coated tissue culture plates to help save time at the bench. Designed to meet a number of different applications, our high quality, consistent Collagen I Coated Plates come in 3 formats – 96 well, 24 well and 6 well plates .
  • Ready to use, room temperature storage
  • Quality tested to help ensure consistency
  • Available in 3 different plate formats and in custom formats for a broad range of applications

    Easy to use, easy to store
    Collagen I, Coated Plates and lids are packed 5 per vacuum sealed foil pouch with a desiccant which can be stored at room temperature for a maximum shelf life of 2 years. Collagen I, rat tail is used to coat tissue culture treated plates. This is the same collagen material we sell in liquid form. The plates are ready to use with no further preparation required, simply seed your cells.

    Quality tested to help ensure consistency
    For maximum efficiency and consistency, Gibco™ Collagen I Coated Plates is designed to meet those expectations. We start with high quality rat tail collagen, the same material available in liquid form and follow a closely controlled, aseptic manufacturing process that produces a very consistent coating on the surface of tissue culture treated plates. We test each lot to help ensure we meet these standards.

    Available in different formats for a broad range of applications
    Extracellular matrix (ECM) proteins play an important role in the regulation of fundamental cellular processes and are widely used as a coating or matrix for tissue culture systems to permit the development of models that closely mimic in vivo conditions. Gibco™ Collagen I Coated Plates can be used in a broad range of applications, including the culturing of fresh or frozen hepatocytes (rat and human), as well as for neuronal, epithelial, and muscle primary cells. They are also ideal for culturing human corneal epithelial cells (HCEC) in defined media.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage 5 plates are sealed in a foil pouch with a desicant for optimal shelf life. Store at room temperature.
    Packaging 5/Pk.
    Product Line Gibco
    Product Type Culture Plate
    No. of Wells 6
    Surface Treatment Cell Culture
    Purity or Quality Grade Cell Culture Grade
    Quantity 5 plates
    Shipping Condition Room Temperature
    Do I need to use all plates from the Collagen I, Coated Plate, 6 well, (Cat. No. A1142801) package at once or can I store the unused plates? If so, for how long can they be stored?

    The unused plates can be resealed in the foil bag and stored for the remaining shelf life of the product.

    What are the dimensions for the Collagen I, Coated Plate, 6 well (Cat. No. A1142801)?

    The dimensions for the Collagen I, Coated Plate, 6 well are as follows:
    - Height: 14.4 mm
    - Length: 127.76 mm
    - Width: 85.48 mm

    What can be used to detach cells from plates when coated with a thin layer of collagen I?

    Enzymes that can be used include: Gibco TrypLE Express Enzyme, Gibco TrypLE Select Enzyme, or Gibco StemPro Accutase Cell Dissociation Reagent.

    What cell types are most commonly cultured in collagen I?

    The following cell types are commonly cultured in collagen I:

    - Endothelial cells (including HUVEC)
    - Fibroblasts
    - Hepatocytes
    - Chondrocytes
    - Osteoclasts
    - Osteoblasts
    - Keratinocytes
    - Corneal epithelium

    How is collagen I used?

    The key applications include the following:

    - Collagen I induces microvascular endothelial cells to adopt a spindle-shaped morphology, in vitro, and to align into solid cord-like assemblies. Vascular endothelial cells can also form vessel-like, tubular structures when cultured on collagen I. Collagen I can be used for in vitro angiogenesis assays.
    - Breast cancer stem cells can undergo differentiation when cultured on collagen I.
    - Collagen I has been used for the culture of primary colon carcinoma cell lines; mouse liver progenitor cells have been cultured in 3D collagen I and rat pancreatic islets.
    - Collagen I can act as a barrier in cell invasion assays, and has been used to study cell adhesion.

    What are some general differences between Gibco Geltrex Matrix, collagen, AlgiMatrix 3D Culture System, and CTS Cellstart Substrate?

    Gibco Geltrex Matrix and collagen (rat tail and bovine) can be used as either a coating solution or a 3D gel matrix. CELLstart Substrate was developed to be used as a xeno-free coating matrix for only ESC applications (as a substitute for Gibco Geltrex Matrix or Matrigel Matrix, which are of animal origin). AlgiMatrix Matrix is a 3D scaffold-type matrix that does not support cell attachment but does provide a good environment for growing spheroids that can be easily harvested for downstream applications.


    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.