Learn More
ILEX LIFE SCIENCES LLC Collagen-I (Type I Collagen) Mouse Native Protein (Tail Tendon), 1 mg
SDP
Catalog No. 502339519
Encompass_Preferred
Native Mouse Collagen-I is a natural protein purified from mouse tail tendon. Purity greater than 90%. FORMULATION: Filtered and lyophilized without additives. OTHER NAMES: Murine Type 1 Collagen, Type I Collagen, Collagen-1. SUMMARY: Collagen, a major extracellular matrix (ECM) component, is a fibrous protein that provides tensile strength to tissues giving them structural integrity. Collagens contribute to mechanical properties, organization, and shape of tissues. Collagen is the most widely used ECM protein for cell culture, facilitating cell attachment, growth, differentiation, migration, and tissue morphogenesis. Type I Collagen is by far the most abundant protein in all vertebrates. It assembles into fibers that form the structural and mechanical scaffold (matrix) of bone, skin, tendons, cornea, blood vessel walls and other connective tissues. Type I collagen consists of two ?1(I) chains and one ?2(I) chain.
Catalog No. 50-233-9519 Supplier ILEX LIFE SCIENCES LLC Supplier No. A126811MG
Native Mouse Collagen-I is a natural protein purified from mouse tail tendon. Purity greater than 90%. FORMULATION: Filtered and lyophilized without additives. OTHER NAMES: Murine Type 1 Collagen, Type I Collagen, Collagen-1. SUMMARY: Collagen, a major extracellular matrix (ECM) component, is a fibrous protein that provides tensile strength to tissues giving them structural integrity. Collagens contribute to mechanical properties, organization, and shape of tissues. Collagen is the most widely used ECM protein for cell culture, facilitating cell attachment, growth, differentiation, migration, and tissue morphogenesis. Type I Collagen is by far the most abundant protein in all vertebrates. It assembles into fibers that form the structural and mechanical scaffold (matrix) of bone, skin, tendons, cornea, blood vessel walls and other connective tissues. Type I collagen consists of two ?1(I) chains and one ?2(I) chain.