Learn More
ILEX LIFE SCIENCES LLC Collagen I Mouse Tail Native Protein 500 ug 500 UG 1/EA
SDP
Catalog No. 5000580177
Encompass_Preferred
Ilex Life Sciences Collagen-I (Type I Collagen) Mouse Native Protein (Tail Tendon) 500 g DESCRIPTION Mouse Collagen-I is a natural protein purified from Mouse tail tendon Collagen-I is purified by proprietary chromatographic techniques Purified Type I Collagen is used in many research applications such as cell culture surface coatings tissue engineering scaffolds 3D bioprinting and cell functional assays FORMULATION Filtered white lyophilized (freeze-dried) powder Collagen-I was lyophilized without additives PURITY 90 0% SHIPPING Ships at ambient temperature STORAGE Lyophilized Collagen-I although stable at room temperature for 3 weeks should be stored desiccated below -18C Upon reconstitution Collagen-I should be stored at 4C between 2-7 days and for future use below -18C For long term storage it is recommended to add a carrier protein (0 1% HSA or BSA) Please prevent freeze-thaw cycles OTHER NAMES Murine Type 1 Collagen Type I Collagen Collagen-1
Catalog No. 50-005-80177 Supplier ILEX LIFE SCIENCES LLC Supplier No. A12681500B
Ilex Life Sciences Collagen-I (Type I Collagen) Mouse Native Protein (Tail Tendon) 500 g DESCRIPTION Mouse Collagen-I is a natural protein purified from Mouse tail tendon Collagen-I is purified by proprietary chromatographic techniques Purified Type I Collagen is used in many research applications such as cell culture surface coatings tissue engineering scaffolds 3D bioprinting and cell functional assays FORMULATION Filtered white lyophilized (freeze-dried) powder Collagen-I was lyophilized without additives PURITY 90 0% SHIPPING Ships at ambient temperature STORAGE Lyophilized Collagen-I although stable at room temperature for 3 weeks should be stored desiccated below -18C Upon reconstitution Collagen-I should be stored at 4C between 2-7 days and for future use below -18C For long term storage it is recommended to add a carrier protein (0 1% HSA or BSA) Please prevent freeze-thaw cycles OTHER NAMES Murine Type 1 Collagen Type I Collagen Collagen-1