Learn More
Enzo Life Sciences Collagen I, rat tail (thin plate coating) (50 ml)
SDP
Catalog No. 501031756
Encompass_Preferred
Alternative name: COL1. MW: 235kDa, 215kDa, 130kDa, 115kDa. Concentration: 100µg/ml. Endotoxin content: <100 EU/mg purified protein (LAL test). Purity: ≥90% (SDS-PAGE). Formulation: Sterile liquid. In 60% ethanol. Appearance: Clear liquid. Source: Isolated from rat tail tendons. Handling: Do not expose to flames. Keep sterile. Do not freeze. Long Term Storage: +4°C
Catalog No. 50-103-1756 Supplier Enzo Life Sciences Supplier No. ALX5224400050
Alternative name: COL1. MW: 235kDa, 215kDa, 130kDa, 115kDa. Concentration: 100µg/ml. Endotoxin content: <100 EU/mg purified protein (LAL test). Purity: ≥90% (SDS-PAGE). Formulation: Sterile liquid. In 60% ethanol. Appearance: Clear liquid. Source: Isolated from rat tail tendons. Handling: Do not expose to flames. Keep sterile. Do not freeze. Long Term Storage: +4°C