Learn More
Medchemexpress LLC Collagen III antibody | 100 UL
SDP
Catalog No. 5000442412
Encompass_Preferred
Collagen III Antibody is a Rabbit-derived and non-conjugated IgG polyclonal antibody, targeting Collagen III. It is intended for research use only.
  • Targets Collagen III
  • Rabbit-derived IgG polyclonal antibody
  • Non-conjugated
  • For research use only
  • WB: 1:500-2000
  • ELISA: 1:5000-10000
  • IHC-P: 1:100-500
  • IHC-F: 1:100-500
  • ICC/IF: 1:100-500
Catalog No. 50-004-42412 Supplier Medchemexpress LLC Supplier No. HYP81101100UL
Collagen III Antibody is a Rabbit-derived and non-conjugated IgG polyclonal antibody, targeting Collagen III. It is intended for research use only.
  • Targets Collagen III
  • Rabbit-derived IgG polyclonal antibody
  • Non-conjugated
  • For research use only
  • WB: 1:500-2000
  • ELISA: 1:5000-10000
  • IHC-P: 1:100-500
  • IHC-F: 1:100-500
  • ICC/IF: 1:100-500