Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Collagen III Antibody | 50 UL
SDP

Catalog No. 5000442518
Encompass_Preferred

Collagen III Antibody is a rabbit-derived and non-conjugated IgG polyclonal antibody that targets Collagen III. It is designed for research use only. Collagen type III is found in most soft connective tissues alongside type I collagen and plays a role in regulating cortical development.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Target: Collagen III
  • Conjugation: Non-conjugated
  • Formulation: Supplied in 0.01M TBS(pH7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles
  • Applications: WB, ELISA, IHC-P, IHC-F, ICC/IF
  • Appearance: Liquid

Catalog No. 50-004-42518 Supplier Medchemexpress LLC Supplier No. HYP8110150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Collagen III Antibody is a rabbit-derived and non-conjugated IgG polyclonal antibody that targets Collagen III. It is designed for research use only. Collagen type III is found in most soft connective tissues alongside type I collagen and plays a role in regulating cortical development.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Target: Collagen III
  • Conjugation: Non-conjugated
  • Formulation: Supplied in 0.01M TBS(pH7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles
  • Applications: WB, ELISA, IHC-P, IHC-F, ICC/IF
  • Appearance: Liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.