Learn More
ILEX LIFE SCIENCES LLC Collagen III Bovine Tissue Native Protein 50 mg 50 MG 1/EA
SDP
Catalog No. 5000580329
Encompass_Preferred
Ilex Life Sciences Collagen-III (Type III Collagen) Bovine Native Protein 50 mg DESCRIPTION Bovine Collagen-III is a natural protein purified from washed dissected bovine tissue into dilute acetic acid Collagen-III is purified by proprietary chromatographic techniques SOURCE Bovine washed dissected tissue FORMULATION Filtered white lyophilized (freeze-dried) powder Collagen-III was lyophilized without additives PURITY 90 0% SHIPPING Ships at ambient temperature STORAGE Lyophilized Collagen-III although stable at room temperature for 3 weeks should be stored desiccated below -18 C Upon reconstitution Collagen-III should be stored at 4 C between 2-7 days and for future use below -18 C For long term storage it is recommended to add a carrier protein (0 1% HSA or BSA) Please prevent freeze-thaw cycles OTHER NAMES Type III Collagen Type 3 Collagen Collagen-3 COL3A1 Collagen Type III Alpha-1 Chain \
Catalog No. 50-005-80329 Supplier ILEX LIFE SCIENCES LLC Supplier No. A1273150
Ilex Life Sciences Collagen-III (Type III Collagen) Bovine Native Protein 50 mg DESCRIPTION Bovine Collagen-III is a natural protein purified from washed dissected bovine tissue into dilute acetic acid Collagen-III is purified by proprietary chromatographic techniques SOURCE Bovine washed dissected tissue FORMULATION Filtered white lyophilized (freeze-dried) powder Collagen-III was lyophilized without additives PURITY 90 0% SHIPPING Ships at ambient temperature STORAGE Lyophilized Collagen-III although stable at room temperature for 3 weeks should be stored desiccated below -18 C Upon reconstitution Collagen-III should be stored at 4 C between 2-7 days and for future use below -18 C For long term storage it is recommended to add a carrier protein (0 1% HSA or BSA) Please prevent freeze-thaw cycles OTHER NAMES Type III Collagen Type 3 Collagen Collagen-3 COL3A1 Collagen Type III Alpha-1 Chain \