Learn More
Diagnostic Biosystems COLLAGEN IV Collagen Type IV
Catalog No. 502407489
Encompass_Preferred
This antibody recognizes collagen type IV epitope located on the α1 and/or α 2 chains of human collagen type IV. This antibody shows no cross-reactivity with collagen types I, II, III, V, VI and VII and does not react with human vitronectin, fibronectin or chondroitin sulfate A, B and C.
Catalog No. 50-240-7489 Supplier Diagnostic Biosystems Supplier No. PDM276
This antibody recognizes collagen type IV epitope located on the α1 and/or α 2 chains of human collagen type IV. This antibody shows no cross-reactivity with collagen types I, II, III, V, VI and VII and does not react with human vitronectin, fibronectin or chondroitin sulfate A, B and C.