Learn More
3HELIX R-CHP (Collagen Hybridizing Peptide, Cy3 Conjugate, 300ug) (RED300)
Catalog No. NC3802725
Encompass_Preferred
The Collagen Hybridizing Peptide (CHP) is a novel probe that specifically targets damaged collagen in vitro and in vivo. By mimicking the Gly-X-Y repeating sequence, it hybridizes with denatured collagen chains to reform the triple helix. This mechanism provides exceptional specificity with minimal non-specific binding. As a powerful histopathology tool, CHP enables straightforward detection of collagen degradation across a wide range of applications. It can be used to visualize tissue damage from inflammation, disease, or aging; track pericellular matrix turnover during cancer cell migration; measure molecular-level mechanical injury; assess collagen in decellularized matrices; and detect collagen bands on SDS-PAGE gels without Western blotting. R-CHP is labeled with sulfo-Cyanine3 for direct fluorescence detection.
Catalog No. NC3802725 Supplier 3HELIX Supplier No. RED300
The Collagen Hybridizing Peptide (CHP) is a novel probe that specifically targets damaged collagen in vitro and in vivo. By mimicking the Gly-X-Y repeating sequence, it hybridizes with denatured collagen chains to reform the triple helix. This mechanism provides exceptional specificity with minimal non-specific binding. As a powerful histopathology tool, CHP enables straightforward detection of collagen degradation across a wide range of applications. It can be used to visualize tissue damage from inflammation, disease, or aging; track pericellular matrix turnover during cancer cell migration; measure molecular-level mechanical injury; assess collagen in decellularized matrices; and detect collagen bands on SDS-PAGE gels without Western blotting. R-CHP is labeled with sulfo-Cyanine3 for direct fluorescence detection.