Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Collagen proline hydroxylase inhibitor | 223666-07-7 | C18H18N4O4 | 100 MG
SDP

Catalog No. 5000374935
Encompass_Preferred

Collagen proline hydroxylase inhibitor is a collagen proline hydroxylase inhibitor, which can be used as an anti-fibrotic agent.

  • Purity: 99.64%
  • Molecular weight: 354.36
  • Appearance: Solid
  • Color: Off-white to light yellow
  • For research use only

Catalog No. 50-003-74935 Supplier Medchemexpress LLC Supplier No. HY15183100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Collagen proline hydroxylase inhibitor is a collagen proline hydroxylase inhibitor, which can be used as an anti-fibrotic agent.

  • Purity: 99.64%
  • Molecular weight: 354.36
  • Appearance: Solid
  • Color: Off-white to light yellow
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.