Learn More
Medchemexpress LLC Collagen proline hydroxylase inhibitor | 223666-07-7 | C18H18N4O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1518350MG
Encompass_Preferred
Collagen proline hydroxylase inhibitor is a chemical compound that functions as an inhibitor of collagen proline hydroxylase. This compound is known for its potential as an anti-fibrotic agent and is often utilized in research related to various disease areas, including cancer.
  • Inhibits collagen proline hydroxylase
  • Functions as an anti-fibrotic agent
  • Used in cancer research (targeted therapy, metabolism, and metastasis)
  • Associated with cytoskeleton and collagen studies
Catalog No. 50-003-72330