Learn More
Crescent Chemical Co Inc Collagen R, 20mL
SDP
Supplier:  Crescent Chemical Co Inc SE47254.01
Encompass_Preferred
Collagen R, 20mL, solution 0.2 % sterile, Type 1 rat tail collagen; 2 mg/ml in 0.1 % acetic acid. Excellent substrate for the culture of hepatocytes, fibroblasts, and epithelial cells. Preparation of collagen film and gels (1, 2). Storage Conditions: +2 °C TO +8 °C
Catalog No. 50-249-116