Learn More
Crescent Chemical Co Inc COLLAGEN R SOLUTION 0.2 %
SDP
Supplier:  Crescent Chemical Co Inc SE47254.02
Encompass_Preferred
Collagen R solution 0.2 % sterile Type 1 rat tail collagen; 2 mg/ml in 0.1 % acetic acid. Excellent substrate for the culture of hepatocytes, fibroblasts, and epithelial cells. Preparation of collagen film and gels (1, 2). Size - 100ML Storage Conditi
Catalog No. 50-249-117