Learn More
Crescent Chemical Co Inc COLLAGEN R SOLUTION 0.4 %
SDP
Catalog No. 50249118
Encompass_Preferred
Collagen R solution 0.4 % sterile Type I rat tail collagen; 4 mg/ml in 0.1 % acetic acid. Excellent substrate for the culture of hepatocytes, fibroblasts, and epithelial cells. Preparation of collagen film and gels (1, 2). Size - 20ML Storage Conditions - +2 °C TO
Catalog No. 50-249-118 Supplier Crescent Chemical Co Inc Supplier No. SE47256.01
Collagen R solution 0.4 % sterile Type I rat tail collagen; 4 mg/ml in 0.1 % acetic acid. Excellent substrate for the culture of hepatocytes, fibroblasts, and epithelial cells. Preparation of collagen film and gels (1, 2). Size - 20ML Storage Conditions - +2 °C TO