Learn More
Ibidi USA Collagen Type I, Rat Tail, 4x5mL
SDP
Catalog No. NC1882578
Encompass
A high-quality rat tail collagen type I solution for cell culture applications (e.g., 3D gels, scaffolds, and coating). Provides in vivo-like extracellular matrix (ECM) structures. Non-pepsinized, native collagen for modelling biological ECM in gel matrices. Fast gelation facilitates optimal cell distribution in 3D gels. Concentration / Volume: 5 mg/ml; 1 x 5 ml Trial Pack
This item is not returnable. View return policy
Catalog No. NC1882578 Supplier Ibidi USA Supplier No. 50205
A high-quality rat tail collagen type I solution for cell culture applications (e.g., 3D gels, scaffolds, and coating). Provides in vivo-like extracellular matrix (ECM) structures. Non-pepsinized, native collagen for modelling biological ECM in gel matrices. Fast gelation facilitates optimal cell distribution in 3D gels. Concentration / Volume: 5 mg/ml; 1 x 5 ml Trial Pack