Learn More
Carbosynth LLC. Affinity purified Rabbit polyclonal Collagen Type I antibody
SDP
Supplier:  Carbosynth LLC. 70RCR007X0.25MG
Encompass_Preferred
Collagen is a group of naturally occurring proteins found, in nature, exclusively in animals, especially in the flesh and connective tissues of mammals. It is the main component of connective tissue, and is the most abundant protein in mammals, making up about 25% to 35% of the whole-body protein content. Type-I collagen is the most abundant collagen of the human body. It is present in scar tissue, the end product when tissue heals by repair. It is found in tendons, the endomysium of myofibrils and the organic part of bone.
Catalog No. 50-125-3427