Learn More
Advanced Biomatrix Rat Tail Collagen Type I Telocollagen 10 mg/ml
Supplier:  Advanced Biomatrix IKD119261001
Encompass_Preferred
Our high concentration rat tail collagen contains 10 mL of 10 mg/ml telocollagen. Being telocollagen, rat tail collagen will form firmer hydrogels than atelocollagen. Our collagen is highly purified, lot-to-lot consistent, and on-the-shelf (available).
Catalog No. 50-225-5949