Learn More
Cell Biologics Collagen Type I, Rat Tail Solution - ready to usm 100ml
SDP
Supplier:  Cell Biologics 6953
Encompass_Preferred
Collagen Type I, Rat Tail Coating Solution (0.1mg/ml) is designed to coat cell culture flasks or dishes. Coat sterile culture dishes or flasks with Collagen Type I, Rat Tail Coating Solution for 2 hours, then aspirate the excess solution and wash dishes with PBS once before seeding cells. The use of coated-dishes improves cell attachment for certain types of primary cells.
Catalog No. NC2644889