Learn More
ILEX LIFE SCIENCES LLC Collagen VI Bovine Placenta Native Protein 1 mg 1 MG 1/EA
SDP
Catalog No. 5000580371
Encompass_Preferred
Ilex Life Sciences Collagen-VI (Type VI Collagen) Bovine Native Protein (Placenta) 1 mg DESCRIPTION Bovine Collagen-VI is a natural protein purified from bovine placenta Collagen-VI is purified by proprietary chromatographic techniques FORMULATION Filtered white lyophilized (freeze-dried) powder Collagen-VI was lyophilized without additives PURITY 90 0% SHIPPING Ships at ambient temperature STORAGE Lyophilized Collagen-VI although stable at room temperature for 3 weeks should be stored desiccated below -18 C Upon reconstitution Collagen-VI should be stored at 4 C between 2-7 days and for future use below -18 C For long term storage it is recommended to add a carrier protein (0 1% HSA or BSA) Please prevent freeze-thaw cycles OTHER NAMES Type VI Collagen Type 6 Collagen Collagen-6 COL6A1
Catalog No. 50-005-80371 Supplier ILEX LIFE SCIENCES LLC Supplier No. A12730100
Ilex Life Sciences Collagen-VI (Type VI Collagen) Bovine Native Protein (Placenta) 1 mg DESCRIPTION Bovine Collagen-VI is a natural protein purified from bovine placenta Collagen-VI is purified by proprietary chromatographic techniques FORMULATION Filtered white lyophilized (freeze-dried) powder Collagen-VI was lyophilized without additives PURITY 90 0% SHIPPING Ships at ambient temperature STORAGE Lyophilized Collagen-VI although stable at room temperature for 3 weeks should be stored desiccated below -18 C Upon reconstitution Collagen-VI should be stored at 4 C between 2-7 days and for future use below -18 C For long term storage it is recommended to add a carrier protein (0 1% HSA or BSA) Please prevent freeze-thaw cycles OTHER NAMES Type VI Collagen Type 6 Collagen Collagen-6 COL6A1