Learn More
Enzo Life Sciences Collagen type XII monoclonal antibody (378D5) (100µl)
SDP
Supplier:  Enzo Life Sciences ALX803309R100
Encompass_Preferred
Clone: 378D5. Isotype: IgG1. Immunogen: Collagen type XII purified from bovine kidney. Species reactivity: Human, Mouse, Rat Chicken, Bovine. Applications: ELISA, IHC (FS), IP, WB. Formulation: Liquid. Ascites. Contains no preservatives.
Catalog No. 50-200-9734