Learn More
Sigma Aldrich Fine Chemicals Biosciences Collagenase from Clostridium histolyticum, 9001-12-1, MFCD00130830, 100 mg
Catalog No. NC0847850
Encompass
For general use, Type I, 0.25-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid, Synonym: Clostridiopeptidase A. Collagenase from Clostridium histolyticum has extensive use in biological studies, where it is used to prepare isolated cell suspensions. This product is suitable for the disaggregation of human tumor, mouse kidney, human adult and fetal brain, lung and many other epithelia tissues.
Catalog No. NC0847850 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C0130100MG
For general use, Type I, 0.25-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid, Synonym: Clostridiopeptidase A. Collagenase from Clostridium histolyticum has extensive use in biological studies, where it is used to prepare isolated cell suspensions. This product is suitable for the disaggregation of human tumor, mouse kidney, human adult and fetal brain, lung and many other epithelia tissues.