Learn More
Worthington Biochemical Corporation COLLAGENASE 4KU CLSPA
SDP
Catalog No. NC9486462
Encompass
Chromatographically purified. <50 caseinase units per milligram. Supplied as a lyophilized powder.
Catalog No. NC9486462 Supplier Worthington Biochemical Corporation Supplier No. LS005275
Chromatographically purified. <50 caseinase units per milligram. Supplied as a lyophilized powder.