Learn More
STEMCELL Technologies Collagenase Type I (0.25%), Size: 5 mL
SDP
Catalog No. NC9952277
Encompass
Collagenase Type I (0.25%) is recommended for tissue dissociation to prepare cell suspensions, dissociation of mouse embryoid bodies (EBs), and solubilization of collagen gels.
Catalog No. NC9952277 Supplier STEMCELL Technologies Supplier No. 07902
Collagenase Type I (0.25%) is recommended for tissue dissociation to prepare cell suspensions, dissociation of mouse embryoid bodies (EBs), and solubilization of collagen gels.