Learn More
AG Scientific Inc Collagenase A Animal Free, 1 G
Catalog No. 502236949
Encompass_Preferred
Collagenase derived from cultures grown in animal-free medium, Suitable for applications needing to avoid introduction of potential animal derived pathogens into bioprocessing procedures.Storage Temperature: +2-8CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6949 Supplier AG Scientific Inc Supplier No. C28451GM
Collagenase derived from cultures grown in animal-free medium, Suitable for applications needing to avoid introduction of potential animal derived pathogens into bioprocessing procedures.Storage Temperature: +2-8CResearch or further manufacturing use only, not for food or drug use.