Learn More
Worthington Biochemical Corporation COLLAGENASE (ANIMAL ORI 1GM
SDP
Catalog No. 50592341
Encompass
Prepared from cultures grown in medium completely devoid of animal based components and designed for bioprocessing applications where introduction of animal derived pathogens must be prevented. 2593-92-3
Catalog No. 50-592-341 Supplier Worthington Biochemical Corporation Supplier No. LS004140
Prepared from cultures grown in medium completely devoid of animal based components and designed for bioprocessing applications where introduction of animal derived pathogens must be prevented. 2593-92-3