Learn More
Sigma Aldrich Fine Chemicals Biosciences Collagenase B from Clostri
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11088831001
Encompass
Collagenase B is prepared from Clostridium histolyticum cultures by filtration ammonium sulfate precipitation dialysis and lyophilization. Collagenase from Clostridium histolyticum is a collagenolytic enzyme. It interacts with collagen-like substances and is responsible for the degradation of collagen into small peptides. It is widely used for the disaggregation of many types of tissues (e.g. lung heart muscle bone adipose tissue liver kidney cartilage mammary gland placentae blood vessels brain tumors) and for the preparation of single-cell suspensions for the establishment of primary cell culture systems. Collagenase B is recommended when yield and viability are important.
Catalog No. 50-291-4558