Learn More
Worthington Biochemical Corporation Collagenase CLSAFA Animal Free, 5 g
SDP
Catalog No. NC1657745
Encompass
CLSAFA is the original AFA grade designed to have collagenase and secondary proteases similar to Types 1 and 2 collagenase. ≥150 units per mg dry weight.
Catalog No. NC1657745 Supplier Worthington Biochemical Corporation Supplier No. LS004156
CLSAFA is the original AFA grade designed to have collagenase and secondary proteases similar to Types 1 and 2 collagenase. ≥150 units per mg dry weight.