Learn More
Sigma Aldrich Fine Chemicals Biosciences Collagenase D from Clostri
Catalog No. 502907355
Encompass
Collagenase D is prepared from Clostridium histolyticum cultures by filtration ammonium sulfate precipitation dialysis and lyophilization.
Catalog No. 50-290-7355 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11088882001
Collagenase D is prepared from Clostridium histolyticum cultures by filtration ammonium sulfate precipitation dialysis and lyophilization.