Learn More
Sigma Aldrich Fine Chemicals Biosciences Collagenase D
Catalog No. NC1578589
Encompass
From Clostridium histolyticum. Collagenase D is prepared from Clostridium histolyticum cultures by filtration, ammonium sulfate precipitation, dialysis, and lyophilization. Collagenase D is recommended when functionality and integrity of cell-surface proteins are important.
Catalog No. NC1578589 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11088866001
From Clostridium histolyticum. Collagenase D is prepared from Clostridium histolyticum cultures by filtration, ammonium sulfate precipitation, dialysis, and lyophilization. Collagenase D is recommended when functionality and integrity of cell-surface proteins are important.