Learn More
Worthington Biochemical Corporation Collagenase/Elastase Vial (CLSH), 5 vials
SDP
Catalog No. NC0449864
Encompass
Enzyme vial of HIS kit that contains 20,000 units of collagenase (Code: CLS-1) and 30 units of elastase (Code: ESL), filtered through 0.22 micron membrane and lyophilized. Store at 2-8°C.
Catalog No. NC0449864 Supplier Worthington Biochemical Corporation Supplier No. LK002067
Enzyme vial of HIS kit that contains 20,000 units of collagenase (Code: CLS-1) and 30 units of elastase (Code: ESL), filtered through 0.22 micron membrane and lyophilized. Store at 2-8°C.