Learn More
Sigma Aldrich Fine Chemicals Biosciences Collagenase from Clostridium histolyticum powder, suitable for cell culture, >=4 FALGPA units/mg solid, high purity, >=700 CDU/mg solid (CDU = collagen digestion units) | 9001-12-1 | MFCD00130830 | 7.5KU
Catalog No. 501780960
Encompass
Collagenase from Clostridium histolyticum powder, suitable for cell culture, >=4 FALGPA units/mg solid, high purity, >=700 CDU/mg solid (CDU = collagen digestion units) | 9001-12-1 | MFCD00130830 | 7.5KU
Catalog No. 50-178-0960 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C27997.5KU
Collagenase from Clostridium histolyticum powder, suitable for cell culture, >=4 FALGPA units/mg solid, high purity, >=700 CDU/mg solid (CDU = collagen digestion units) | 9001-12-1 | MFCD00130830 | 7.5KU