Learn More
Sigma Aldrich Fine Chemicals Biosciences Collagenase from Clostridium histolyticum release of physiologically active rat pancreatic islets tested, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid | 9001-12-1 | MFCD00130830 | 25mg
Catalog No. 501780973
Encompass
Collagenase from Clostridium histolyticum release of physiologically active rat pancreatic islets tested, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid | 9001-12-1 | MFCD00130830 | 25mg
Catalog No. 50-178-0973 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C765725MG
Collagenase from Clostridium histolyticum release of physiologically active rat pancreatic islets tested, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid | 9001-12-1 | MFCD00130830 | 25mg