Learn More
Worthington Biochemical Corporation COLLAGENASE III 1GCOLLAGENASE
SDP
Catalog No. NC9405360
Encompass
Collagenase; Type 3; 1g; Secondary proteolytic contaminant activities but with typical collagenase activity; Minimum Activity: > /=100U/mg dry weight; Source: Clostridium histolyticum; A dialyzed, lyophilized powder Stored at 2 deg. to 8 deg.C
Catalog No. NC9405360 Supplier Worthington Biochemical Corporation Supplier No. LS004182
Collagenase; Type 3; 1g; Secondary proteolytic contaminant activities but with typical collagenase activity; Minimum Activity: > /=100U/mg dry weight; Source: Clostridium histolyticum; A dialyzed, lyophilized powder Stored at 2 deg. to 8 deg.C