Learn More
Advanced Biomatrix Collagenase, Lyophilized Powder, 50 mg
Catalog No. NC1668831
Encompass_Preferred
This Collagenase product is provided as a lyophilized, sterile powder in a 50 mg package size. It has been tested specifically and shown to effectively hydrolyze collagen and dissociate tissues. Collagenase consists of a blend of enzymes including collagenase, caseinase, clostripain and trypsin designed to effectively hydrolyze collagen and dissociate tissues. Various collagenase products have been evaluated specifically for use with Advanced BioMatrix’ s collagen products. This product is offered to improve the viability and functionality of isolated cells from the digested collagen gels. After reconstitution, the product is ready-to-use.
Catalog No. NC1668831 Supplier Advanced Biomatrix Supplier No. 503050MG
This Collagenase product is provided as a lyophilized, sterile powder in a 50 mg package size. It has been tested specifically and shown to effectively hydrolyze collagen and dissociate tissues. Collagenase consists of a blend of enzymes including collagenase, caseinase, clostripain and trypsin designed to effectively hydrolyze collagen and dissociate tissues. Various collagenase products have been evaluated specifically for use with Advanced BioMatrix’ s collagen products. This product is offered to improve the viability and functionality of isolated cells from the digested collagen gels. After reconstitution, the product is ready-to-use.