Learn More
Medchemexpress LLC Collagenase/neutral protease 2 (animal free) | 10 MG
SDP
Catalog No. 5000296962
Encompass_Preferred
An animal-free enzymatic mixture of collagenase and neutral protease for tissue dissociation and primary cell isolation. Supplied as a light yellow to brown solid for research use; store refrigerated at 4°C. Comes with quality and safety documentation for reproducible cell dissociation protocols.
  • Animal-free collagenase and neutral protease mixture for tissue dissociation.
  • Formulated for primary cell and stem cell isolation.
  • Solid formulation, suitable for reconstitution in buffer for protocols.
  • Stable when stored at 4°C; shelf life indicated as 2 years on COA.
  • Includes COA and SDS documentation to support batch-level quality control.
Catalog No. 50-002-96962 Supplier Medchemexpress LLC Supplier No. HYE7053110MG
An animal-free enzymatic mixture of collagenase and neutral protease for tissue dissociation and primary cell isolation. Supplied as a light yellow to brown solid for research use; store refrigerated at 4°C. Comes with quality and safety documentation for reproducible cell dissociation protocols.
  • Animal-free collagenase and neutral protease mixture for tissue dissociation.
  • Formulated for primary cell and stem cell isolation.
  • Solid formulation, suitable for reconstitution in buffer for protocols.
  • Stable when stored at 4°C; shelf life indicated as 2 years on COA.
  • Includes COA and SDS documentation to support batch-level quality control.