Learn More
Sigma Aldrich Fine Chemicals Biosciences Collagenase P from Clostri
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11213857001
Encompass
Collagenase P is an enzyme mixture prepared from the extracellular culture filtrate of a special Clostridium histolyticum strain.Collagenase P is a research biochemical not manufactured under the US good manufacturing practices (GMP) regulations.Note Collagenase P is a non-sterile preparation of the culture supernatant of Clostridium histolyticum. It is neither designed nor intended for isolation of pancreatic islets for transplantation into humans and must not be used for such purposes.
Catalog No. 50-290-0078