Learn More
Worthington Biochemical Corporation COLLAGENASE TYPE 3 100MG
SDP
Supplier:  Worthington Biochemical Corporation LS004180
Encompass
Lower in secondary proteolytic contaminant activities but with typical collagenase activity. Suggested for mammary primary cell isolation. A dialyzed, lyophilized powder.
Catalog No. NC9449488