Learn More
Worthington Biochemical Corporation Collagenase, Type 2, 1g
SDP
Supplier:  Worthington Biochemical Corporation LS004176
Encompass
Prepared to contain higher clostripain activity. Suggested for bone, heart, liver, thyroid and salivary primary cell isolation. Supplied as a dialyzed, lyophilized powder. Store at 2-8°C.
Catalog No. NC9693955