Learn More
STEMCELL Technologies Collagenase Type IV, Size: 5 g
SDP
Supplier:  STEMCELL Technologies 1000680
Encompass
Collagenase is a protease consisting of a single polypeptide chain approximately 1000 amino acid residues in length. Collagenase is capable of digesting native collagen fibrils commonly found in connective tissues and therefore is frequently used for tissue dissociation. Collagenase preparations contain the activity of several proteases including collagenase caseinase clostripain and trypsin (Kessler Yaron). Collagenase Type IV contains low levels of tryptic activity and is frequently used for pancreatic islets (Taguchi et al.) and for applications where maintenance of receptor integrity is required.
Catalog No. 50-239-0194