Learn More
STEMCELL Technologies Collagenase Type V, Size: 100 mg
SDP
Supplier:  STEMCELL Technologies 07430
Encompass
Collagenase is a protease consisting of a single polypeptide chain approximately 1,000 amino acid residues in length. Collagenase is capable of digesting native collagen fibrils commonly found in connective tissues and therefore is frequently used for tissue dissociation. Collagenase preparations contain the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin (Kessler & Yaron). Collagenase Type V contains high levels of collagenase and caseinase activities.
Catalog No. 50-197-4398