Learn More
Worthington Biochemical Corporation COLLAGENESE TYPE 1 FILTERED
SDP
Supplier:  Worthington Biochemical Corporation LS004214
Encompass
Collagenase, Type 1 (Code: CLS-1), Filtered Through a 0.22 Micron Membrane and Lyophilized in Vials to Contain 50mg per Vial, Store at 2 deg. to 8 deg.C, Source: Clostridium histolyticum, Minimum Activity: 125 units/mg Dry Weight, 50mg
Catalog No. NC9068181